LaMotte® Turbidity Test Kit
Catalog #: 470227-856
Supplier: LaMotte
CAS Number:
Specifications
- Description:Turbidity Test Kit
- Size:1 Kit
- Cat. no.:470227-856
- No. of Tests:100
Specifications
About this item
This kit tests for turbidity using a turbidmetric card used match clarity to your sample.
- Unlimited tests
Range and sensitivity is 0, 20, 40, 60, 80, 100 JTU.
Customers Who Bought This Also Bought