To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Rabbit Complement
- Size:1 ml
- Storage temperature:Store at 2-8°C.
- Application:Immunoassays
- Cat. no.:IC0855866
Specifications
About this item
Product is rabbit serum, rapidly processed to preserve complement activity. Pooled rabbit serum is filtered through a 0.22 µm
- This product is suitable for use in hemolytic titrations and cytotoxicity assays, and as a serum protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques.
- This product has not been absorbed, and should be tested for use in specific cytotoxicity applications.
The product gives a spectrum of normal serum precipitin arcs when tested by immunoelectrophoresis with anti-rabbit serum. The product is tested for its ability to lyse red blood cells.