Specifications
- Source:Pokeweed (Phytolacca americana)
- Size:1 mg
- Storage conditions:Store lyophilized product at 2-8 °C. After reconstitution, aliquot and store at –20 °C. Avoid Freeze/Thaw cycles.
- Biological activity:0.03-0.3 μg/mL mitogenic activity
- Protein/peptide name:Mitogen
- Solubility:Soluble in phosphate buffered saline (1 mg/mL).
- Formulation:Lyophilized
- Tested applications:Immunoassays
- Cat. no.:IC08780341
Specifications
About this item
Lectin is pure Phytolacca americana from Pokeweed.
- Mitogenic assay
- Extinction coefficient: E1% = 18.5 (280 nm in water) (Lit.)
- UV/Vis Absorbance: λ max: 280 nm (Lit.)
- MDL No: MFCD00161973
- Pure pokeweed mitogen is used in mitogenic assays. It has variety of applications in vitro like blood grouping and erythrocyte polyagglutination studies, mitogenic stimulation of lymphocytes, lymphocyte subpopulation studies, fractionation of cells and other particles, histochemical studies of normal and pathological conditions.
Lectins are proteins or glycoproteins of non-immune origin that agglutinate cells and/or precipitate complex carbohydrates. Lectins are capable of binding glycoproteins even in presence of various detergents. The agglutination activity of these highly specific carbohydrate-binding molecules is usually inhibited by a simple monosaccharide, but for some lectins, di, tri, and even polysaccharides are required.
Pokeweed mitogen has specificity for sugar chains containing a di-N-Acetylchitobiose moiety such as is found on "band-3 glycoprotein" from human erythrocytes. Di-N-acetylchitobiose and tri-N-acetylchitobiose inhibit agglutination activity of the lectin.
PWM is extracted from pokeweed roots. This protein has hemagglutinating, leukoagglutinating, and mitogenic properties. PWM is not blood group specific, but has affinity for N-acetyl-β-D-glucosamine oligomers.
Reconstitution: Reconstitute with 0.01M sodium phosphate-0.15M sodium chloride, pH 7.2-7.4.