To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Source:Human
- Species:Human
- Size:5 mg
- Storage conditions:Store unopened product at 2-8 °C. Store reconstituted material no longer than a few days at 2-8 °C unless sodium azide is added. For long-term storage, aliquot and freeze at –20 °C
- Protein/peptide name:IgA
- Purity:>99%
- Concentration:Total Protein: 5.41 mg/mL; Volume: 1 mL
- Formulation:Lyophilized powder
- Tested applications:Immunoassays
- Cat. no.:IC0855906
Specifications
About this item
Product is the lyophilized powder of human serum IgA and buffer salts.
- Human IgA is purified from pooled human serum using multi-step procedures which may include salt fractionation, gel filtration, ion-exchange chromatography and immunoabsorption.
- The product is dialyzed into 0.01M sodium phosphate, 0.07M sodium chloride, pH 7.3, filtered through a 0.22 µm filter, vialed and lyophilized.
- No preservative has been added.
- This product is suitable for use as a positive control in antibody identification screening techniques and is recommended as an
- ELISA coating antigen and control.
- Purified human serum IgA is suitable for use as a positive control in antibody identification screening techniques and is recommended as an ELISA coating antigen and control.
- From human plasma highly purified, immunoabsorbed to remove trace amounts of contaminant proteins
The product is tested for purity at a minimum of 20 mg/mL using immunoelectrophoresis. The product is human serum IgA; no trace of IgG, IgM or non-immunoglobulin serum proteins is detected. The total protein is determined using the Biuret procedure, with bovine albumin as the standard. Each vial is overfilled by approximately 10 percent so that a minimum of 5 mg of human serum IgA is obtained upon reconstitution.
Human IgA is purified from pooled human serum using multi-step procedures which may include salt fractionation, gel filtration, ion-exchange chromatography and immunoabsorption. The product is dialyzed into 0.01M sodium phosphate, 0.07M sodium chloride, pH 7.3, filtered through a 0.22 μm filter, vialed and lyophilized.