Specifications
- Packaging:Plastic Ampoule
- Size:1 mg
- Protein/peptide name:Angiotensin
- Cat. no.:CA80011-516
- Supplier No.:05-23-0101-1MG
Specifications
About this item
Angiotensin II plays an important role in the regulation of blood pressure.
- Peptide sequence: H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-OH
Angiotensin II stimulates the release of aldosterone from the adrenal gland. Inhibits adenylate cyclase activity in spontaneously hypertensive rats. Has strong vasoconstrictive effects. Increases the entry of Ca²⁺ in heart muscle via voltage-sensitive channels and activates myosin light chain kinase. Also elicits an increase in voltage-dependent delayed rectifier K⁺ current in neurons. Activates JAK2 in smooth muscle cells. Activates p125FAK and a cytosolic 115 to 120 kDa calcium-dependent tyrosine kinase in rat epithelial cells. Also activates p60c-src as well as ERK1/2, JNK, and p38MAP Kinase in vascular smooth muscle cells.