Magnesium acetate tetrahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog #: CAJT2425-7
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 16674-78-5
MOC: Non-Regulated
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PE pail, screw top
- Size:12 kg
- Cat. no.:CAJT2425-7
- Supplier No.:2425-07
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay ((CH₃COO)₂Mg·4H₂O):98.0 - 102.0 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - pH of 5% Solution at 25°C:6.5 - 8.5
- ACS - Chloride (Cl):≤0.001 %
- ACS - Sulfate (SO₄):≤0.005 %
- ACS - Barium (Ba):≤0.001 %
- ACS - Calcium (Ca):≤0.01 %
- ACS - Manganese (Mn):≤0.001 %
- ACS - Potassium (K):≤0.005 %
- ACS - Sodium (Na):≤0.005 %
- ACS - Strontium (Sr):≤0.005 %
- ACS - Endotoxin Concentration (2.5 EU/g max.):Passes Test
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- J.T.Baker®, Magnesium acetate tetrahydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS:
Specification Test Results
About this item
Synonyms: Acetic acid magnesium salt tetrahydrate
Density:1.45 g/cm³
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-102,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 80 °C
:Flash Pt:
:Flash Pt:
:M.W. 214.46 g/mol
:80
:M.O.C. Category N
Formula:(H₃CCOO)₂Mg·4H₂O
MW:214.46 g/mol
MDL Number:MFCD00149214
CAS Number:16674-78-5
Customers Who Bought This Also Bought