Silica gel 60, SilicAR (230-400 Mesh) for column chromatography (60A 40-63 µm), Macron Fine Chemicals™
Supplier: Avantor
MOC: Non-Regulated
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MW:60.08 g/mol
MDL Number:MFCD00011232
:Length:
:Width:
:Height:
:Ø
:Thickness:
:(230-400 Mesh)
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 1710 °C
:Flash Pt:
:Flash Pt:
:M.W. 60.08 g/mol
:Silica gel
:1710
:2230
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Appearance (white powder):Passes Test
- Specific Surface Area, m²/g:450 - 550
- Pore Volume (ml/g):0.65 - 0.85
- Mean Pore Diameter, A:50 - 70
- <25 um:≤15 %
- >71 um:≤7 %
- pH:6.0 - 7.5
- Water Content:≤9 %
- Macron Fine Chemicals™, Silica gel 60, SilicAR (230-400 Mesh) for column chromatography (60A 40-63 µm):
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_MACRV120
SDS
V120 Product Specifications
Additional Product Details
Customers Who Bought This Also Bought