TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0% (dried basis), Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Catalog #: CAJT4108-07
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 1185-53-1
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PE pail, screw top
- Size:12 kg
- Cat. no.:CAJT4108-07
Specifications
Specification Test Results
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Assay (NH₂C(CH₂OH)₃·HCl) (dried basis):≥99.0 %
- Appearance:Passes Test
- Identification (by IR):Passes Test
- Absorbance of a 1M Solution - 260 nm:≤0.06
- Absorbance of a 1M Solution - 280 nm:≤0.06
- Absorbance of a 1M Solution - 400 nm:≤0.01
- Water (H₂O)(by Karl Fischer titrn):≤0.5 %
- RNase Activity (None Detected):Passes Test
- DNase Activity (None Detected):Passes Test
- Protease Activity (None Detected):Passes Test
- Trace Impurities - Calcium (Ca):≤1 ppm
- Trace Impurities - Copper (Cu):≤1 ppm
- Trace Impurities - Iron (Fe):≤1 ppm
- Trace Impurities - Lead (Pb):≤1 ppm
- Trace Impurities - Magnesium (Mg):≤1.000 ppm
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Manufactured exclusively for Avantor Performance Materials, LLC., by:
- BioSpectra, Inc., Stroudsburg, PA:
Specification Test Results
About this item
Synonyms: Tris(hydroxymethyl)methylamine hydrochloride,2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride,Trizma(R) hydrochloride,Tris(hydroxymethyl)aminomethane hydrochloride,TRIS HCl,Tris(hydroxymethyl)methylammonium chloride,Trometamol hydrochloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 150 to 152 °C
:Flash Pt:
:Flash Pt:
:M.W. 157.6 g/mol
:TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride)
:GHS07
:M.O.C. Category R
:12,09902
Formula:NH₂C(CH₂OH)₃·HCl
MW:157.6 g/mol
MDL Number:MFCD00012590
CAS Number:1185-53-1
Customers Who Bought This Also Bought