J.T.Baker® Cycle-Tap, Dispenser System
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
The cycle-tap dispenser is a stainless steel solvent dispense unit with a hose designed for unmetered manual dispensing of solvent.
- Fully assembled with color-coded quick-connect
- Three-way valve for purging to ensure highest purity
- Stainless-steel components that are inert to solvents to maintain solvent quality
- Safety armored, flexible hose with internal steel braiding that will not kink or puncture
- Thumb operated toggle valve with 'hold-on' option for easy dispensing
- Integrated hook for hanging the dispenser’s outlet
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.