Kilo Scale Add-On Distillation Kit, Ace Glass Incorporated
Catalog #: 76291-814
Supplier: Ace Glass
Specifications
- Description:Kilo-Scale Add-On Distillation Kit
- Cat. no.:76291-814
Specifications
About this item
Add-on distillation kit for use with the ACE Universal Stand as part of any ACE kilo scale reactor. Kit includes a side shelf to attach to an existing stand, a receiving flask, a distillation splitter, a condenser, tubing, and adapters.
Customers Who Bought This Also Bought