Potassium sulfate 99.0-100.5% FCC, purified, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Formula:K₂SO₄
MW:174.26 g/mol
Boiling Pt:1698 °C (1013 hPa)
Melting Pt:1069 °C
Density:2.66 g/cm³ (20 °C)
MDL Number:MFCD00011388
CAS Number:7778-80-5
Merck Index:12,07845
:Storage Temperature: Ambient
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = RL
:Boiling Pt:
:Boiling Pt: 1698 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 1069 °C
:Flash Pt:
:Flash Pt:
:M.W. 174.26 g/mol
:Potassium sulfate
EINECS:231-915-5
:1698 °C (1013 hPa)
Storage Temperature:Ambient
:M.O.C. Category RL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets F.C.C. Requirements:
- Food GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Assay (K₂SO₄):99.0 - 100.5 %
- Appearance (white powder):Passes Test
- Identification A:Passes Test
- Identification B:Passes Test
- Solubility:Passes Test
- Heavy Metals (as Pb):≤10 ppm
- Lead (Pb):≤2 mg/kg
- Selenium (Se):≤5 mg/kg
- pH:5.5 - 8.5
- Reaction:Passes Test
- Chloride (Cl):≤0.07 %
- Arsenic (As):≤3 ppm
- Calcium (Ca):Passes Test
- Magnesium (Mg):≤0.1 %
- Sodium (Na):≤0.05 %
- On U.S. No. 80 Sieve:≤5 %
- On U.S. No. 120 Sieve, For Information Only:
- Macron Fine Chemicals™, Potassium sulfate 99.0-100.5% FCC, purified:
Specification Test Results
Customers Who Bought This Also Bought