Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis) NF, ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Soda,Carbonic acid sodium salt,Disodium carbonate,Na-X,Bisodium carbonate,Carbonic acid, disodium salt
Formula:Na₂CO₃
MW:105.99 g/mol
MDL Number:MFCD00003494
CAS Number:497-19-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 854 °C
:Flash Pt:
:Flash Pt:
:M.W. 105.99 g/mol
:854
:1600
:GHS07
:12,08739
:14,08596
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets N.F. Requirements:
- Meets ACS Reagent Chemical Requirements:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Water (H₂O):≤0.5 %
- NF - Assay (Na₂CO₃) (anhydrous basis):99.5 - 100.5 %
- ACS - Assay (Na₂CO₃) (dried basis)(by acidimetry):≥99.5 %
- ACS - Calcium (Ca):≤0.03 %
- ACS - Chloride (Cl):≤0.001 %
- ACS - Heavy Metals (as Pb):≤5 ppm
- ACS - Insoluble Matter:≤0.01 %
- ACS - Iron (Fe):≤5 ppm
- ACS - Loss on Heating at 285°C:≤1.0 %
- ACS - Magnesium (Mg):≤0.005 %
- ACS - Phosphate (PO₄):≤0.001 %
- ACS - Potassium (K):≤0.005 %
- ACS - Silica (SiO₂):≤0.005 %
- ACS - Sulfur Compounds (as SO₄):≤0.003 %
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
- J.T.Baker®, Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis) NF, ACS:
Specification Test Results
Customers Who Bought This Also Bought