About this item
HyClone™ Characterized FBS (New Zealand origin) is widely used by users requiring an extensive biochemical profile and full traceability.
- Sourced in New Zealand with complete traceability back to the original source
- Low in antibodies and high in growth factors
- Produced using true pool technology for uniformity and consistency from bottle to bottle
- Filtered through three sequential 100 nm (0,1 µm) pore size-rated filters
- Virus panel testing according to 9 CFR 113,53
HyClone Characterised Fetal Bovine Serum (FBS), New Zealand Origin is carefully collected, processed, and filtered in our own New Zealand facility. The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically. One of the benefits of this isolation is that New Zealand has the fewest reported bovine diseases in the world, making Characterised FBS, New Zealand Origin an excellent source of bovine serum as it offers greater security.
Characterised FBS is assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase, glutamic oxaloacetic transaminase, pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC), percent iron saturation, glucose, and IgG. Included assays are subject to change without notice.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA76324-906
- CA76324-908
- CA76324-938
- CA76236-356
- CA95059-628
- Origin:
- Animal
- Animal
- Animal
- Animal
- Animal
- Endotoxin:
- ≤20 EU/ml
- ≤20 EU/ml
- ≤20 EU/ml
- ≤20 EU/ml
- ≤20 EU/ml
- Hemoglobin:
- <25 mg/dl
- <25 mg/dl
- <25 mg/dl
- <25 mg/dl
- <25 mg/dl
- Storage temperature:
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
Specifications
Related Information
Certificates
Search by Lot NumberCytiva BioXtra and Pall Life Sciences Products CA Cytiva BioXtra and Pall Life Sciences Products US
Promotion Material
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Cytiva HyClone Media and Supplements HyClone™ Cell Culture Serum products