Order Entry
ContactUsLinkComponent
Anti-TP53 Rabbit Polyclonal Antibody
Anti-TP53 Rabbit Polyclonal Antibody
Catalog #: 76262-724
P53 K292 Ac PAB 25UL
Anti-TP53 Rabbit Polyclonal Antibody
Catalog #: 76262-724
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    TP53
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Modification:
    Acetylation
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-724

Specifications

About this item

TP53 (tumor suppressor gene p53) is one of the most well-studied genes that suppresses tumor formation and renders protection against DNA damage by inducing cell cycle arrest, DNA repair, or apoptosis. TP53 signaling is triggered through numerous cellular events ranging from DNA damage to hypoxia, stress and a plethora of other causes. Upon activation, p53 acts as zinc-containing transcriptional regulator and initiates a cascade of events that determines the cellular outcome including cell cycle arrest, apoptosis, senescence, DNA repair, development, differentiation and tissue homeostasis. Cell cycle arrest is induced by p53 via trans-activating genes such as p21 (CDK-inhibitor 1, cyclin dependent kinase) and others. Interestingly, p53 itself is capable of triggering cellular responses (survival or induced cell death) as well. Mutations or deletions in the TP53 gene are present in nearly 50% of human cancers, and primarily results in impaired tumor suppressor function. Anti-p53 (ac Lys292) antibody is ideal for researchers interested in developmental biology, cell growth and cancer research.

p53 K292 Ac Antibody is suitable for ELISA and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~43.6 kDa in size corresponding to p53 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: TP53
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought