Order Entry
ContactUsLinkComponent
Anti-TJP1 Rabbit Polyclonal Antibody
Anti-TJP1 Rabbit Polyclonal Antibody
Catalog #: 76262-714
ZO-1 Polyclonal Antibody 100UG
Anti-TJP1 Rabbit Polyclonal Antibody
Catalog #: 76262-714
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    TJP1
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-714

Specifications

About this item

ZO-1, also called TJP1, belongs to the MAGUK family. This gene encodes a protein located on a cytoplasmic membrane surface of intercellular tight junctions. The encoded protein may be involved in signal transduction at cell-cell junctions. The N-terminal may be involved in transducing a signal required for tight junction assembly, while the C-terminal may have specific properties of tight junctions. The alpha domain might be involved in stabilizing junctions. ZO-1 plays a role in the regulation of cell migration by targeting CDC42BPB to the leading edge of migrating cells. ZO1 may be associated the following disorders; celiac desease, congenital nephrotic syndrome finnish type, and macular degeneration. Anti-ZO-1 Antibody is useful for researchers interested in Apoptosis Research and Insulin Research.

Anti-ZO-1 Antibody has been tested in Western Blot, ELISA, Immuohistochemistsry, Immunofluorescence, and Flow Cytometry. Expect a band at ~245 and/or 195.5 kDa in western blot using appropriate lysates. Positive control whole cell lysates used A549 and PC3 @ 1µg/mL for WB; CACO2 and PC3 with PFA and MeOH @ 10µg/mL for IF. Positive control cells for FC were PC3 and positive control tissues for IHC was mouse adipose tissue.

Type: Primary
Antigen: TJP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought