Order Entry
ContactUsLinkComponent
Anti-TAF10 Rabbit Polyclonal Antibody
Anti-TAF10 Rabbit Polyclonal Antibody
Catalog #: 76262-964
TAF10 Polyclonal Antibody 25UL
Anti-TAF10 Rabbit Polyclonal Antibody
Catalog #: 76262-964
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    TAF10
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-964

Specifications

About this item

TAF10 (TATA-Box Binding Protein Associated Factor 10) is the protein that coordinates activities for the initiation of transcription by RNA polymerase II required for activities of more than 70 polypeptides. TFIID binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as co-activators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the small subunits of TFIID that is associated with a subset of TFIID complexes. Studies with human and mammalian cells have shown that this subunit is required for transcriptional activation by the estrogen receptor, for progression through the cell cycle, and may also be required for certain cellular differentiation programs. Anti-TAF10 Antibody is useful for researchers interested in Chromatin research, transcription factor activity research, and DNA binding and transcription co-activator activity research.

Anti-TAF10 Antibody has been tested for use in ELISA, Dot Blot, and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 21.7 kDa in size corresponding to TAF10 protein by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: TAF10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought