Order Entry
ContactUsLinkComponent
Anti-STAT3 Rabbit Polyclonal Antibody
Anti-STAT3 Rabbit Polyclonal Antibody
Catalog #: 76263-042
STAT3 R31-Me2a PAB 25UL
Anti-STAT3 Rabbit Polyclonal Antibody
Catalog #: 76263-042
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    STAT3
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Modification:
    Methylation
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76263-042

Specifications

About this item

Signal transducer and activator of transcription 3 (Stat3) belongs to a family of cytoplasmic transcription factors that can be activated by phosphorylation by its cell surface receptor. Stat3 plays a key role in many cellular processes such as cell growth and apoptosis. It also mediates cellular responses to interleukins, KITLG/SCF,EGF, IFN-alpha and other growth factors and may mediate cellular responses to activated FGFR1, FGFR2, FGFR3 and FGFR4. Stat3 forms a homodimer or a heterodimer with a related family member (e.g. STAT1). Activation occurs through phosphorylation of tyrosine 705 and serine 727. Phosphorylation of Stat3 at Tyr705 induces Stat3 dimerization and nuclear translocation. Serine phosphorylation is important for stable DNA-binding of Stat3 homodimers and maximal transcriptional activity. Stat3 can have a dual role in cancer, it has been found that Stat3 protein can promote oncogenesis and have a tumor suppressor role depending upon the mutational background of the tumor.

Anti-STAT3 [Asym-dimethyl Arg31] antibody is useful for Western Blot and Dot Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~88 kDa corresponding to STAT3 protein by Western Blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: STAT3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought