Order Entry
ContactUsLinkComponent
Anti-SIRT1 Rabbit Polyclonal Antibody
Anti-SIRT1 Rabbit Polyclonal Antibody
Catalog #: 76262-744
SIRT1 Polyclonal Antibody 25UL
Anti-SIRT1 Rabbit Polyclonal Antibody
Catalog #: 76262-744
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    SIRT1
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-744

Specifications

About this item

SIRT1 antibody detects human SIRT1 protein. SIRT1 is a member of the sirtuin family of protein-modifying enzymes. SIRT1 is a NAD+-dependent deacetylase that plays an important role in many cellular processes. SIRT1 protein is known to regulate epigenetic gene silencing and suppress recombination of rDNA. SIRT1 deacetylates a wide range of substrates, including p53, NF-kB, FOXO transcription factors, and PGC-1 alpha, with roles in cellular processes such as muscle differentiation, adipogenesis, protection from axonal degeneration, and life span extension. SIRT1 is downregulated in cells that have high insulin resistance and inducing its expression increases insulin sensitivity, suggesting the molecule is associated with improving insulin sensitivity. Furthermore, SIRT1 de-acetylates and affects the activity of both members of the PGC1-alpha/ERR-alpha complex, which are essential metabolic regulatory transcription factors. Anti-SIRT1 Antibody is useful for researcher involved in research areas including cancer, diabetes, aging, neurodegenerative, metabolic or cardiovascular diseases.

This affinity purified antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band ~62 or 81.7 kDa in size corresponding to SIRT1 by western blotting in the appropriate cell lysate or extract. Higher observed band ~125 kda is due to complexing proteins.

Type: Primary
Antigen: SIRT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought