Order Entry
ContactUsLinkComponent
Anti-Rorc Rabbit Polyclonal Antibody
Anti-Rorc Rabbit Polyclonal Antibody
Catalog #: 76263-048
Nuclear receptor ROR gamma pS203 Polyclonal Antibody 25UL
Anti-Rorc Rabbit Polyclonal Antibody
Catalog #: 76263-048
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    Rorc
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Modification:
    Phosphorylation
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76263-048

Specifications

About this item

Anti-Nuclear receptor ROR gamma pS203 was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Nuclear receptor ROR gamma pS203 antibody detects mouse receptor ROR gamma phosphorylated at the serine 203 position. RAR-related orphan receptor gamma is a member of the nuclear receptor family of transcription factors. (ROR-gamma) is a key regulator of cellular differentiation, immunity, peripheral circadian rhythm as well as lipid, steroid, xenobiotics and glucose metabolism. ROR-gamma regulates the circadian expression of clock genes such as CRY1, ARNTL/BMAL1 and NR1D1 in peripheral tissues and in a tissue-selective manner. It is also involved in the regulation of the rhythmic expression of genes involved in glucose and lipid metabolism and is a negative regulator of adipocyte differentiation. It controls adipogenesis as well as adipocyte size and modulates insulin sensitivity in obesity. Isoform 2, ROR-gamma is essential for lymphoid organogenesis, in particular lymph nodes and Peyer's patches. ROR-gamma also plays an important regulatory role in thymopoiesis, and in inhibiting apoptosis of undifferentiated T cells. Anti-nuclear receptor ROR gamma pS203 is ideal for researcher's interested in autoimmune diseases such as psoriasis and rheumatoid arthritis, circadian rhythms, and immune system disorders.

This affinity purified antibody has been tested for use in ELISA and western blot.   Specific conditions for reactivity should be optimized by the end user. Expect a band ~ 61 kDa in size corresponding to receptor ROR gamma by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: Rorc
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought