Order Entry
ContactUsLinkComponent
Anti-Ngp Rabbit Polyclonal Antibody
Anti-Ngp Rabbit Polyclonal Antibody
Catalog #: 76262-746
Ngp Polyclonal Antibody 100UG
Anti-Ngp Rabbit Polyclonal Antibody
Catalog #: 76262-746
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Rabbit
  • Antigen symbol:
    Ngp
  • ImmunoFluorescence:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-746

Specifications

About this item

This antibody is designed, produced, and validated as part of a collaboration between Rockland and the National Cancer Institute (NCI). A proteomic study in 2011 determined a direct correlation between NGP mRNA levels and myeloid-derived suppressor cells. Specifically cathepsin B, a catalytic mechanism and potentially biomarker in a variety of cancers and amyloid plaque development, was inhibited by NGP. The significance of NGP as a negative regulator of tumor metastasis makes this antibody ideal for investigators involved in cancer research, as well as Alzheimer’s research.

This affinity purified antibody has been tested for use in IF, and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band 39-42 kDa for non reduced and 20-27 kDa for reduced gels corresponding to Ngp by western blotting in the appropriate cell lysate or extract. 

Type: Primary
Antigen: Ngp
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: rabbit

Customers Who Bought This Also Bought