Order Entry
ContactUsLinkComponent
Anti-MAPK3 Rabbit Polyclonal Antibody (FITC)
Anti-MAPK3 Rabbit Polyclonal Antibody (FITC)
Catalog #: 76262-794
Anti-MAPK3 Rabbit Polyclonal Antibody (FITC)
Catalog #: 76262-794
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    MAPK3
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Ambient
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-794

Specifications

About this item

ERK1 antibodies detect the ERK1 isoform. Mitogen activiated protein kinase 3, also known as MAPK3, ERK, or ERK1, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. ERK1 and ERK2 are activated by MEK1 and MEK2 in the B-raf signaling pathway resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Human ERK1 and ERK2 are 84% identical in sequence and share common functionality in cells. Anti-ERK1 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-ERK 1 (RABBIT) antibody is suitable for use in ELISA and Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 43 kDa.

Type: Primary
Antigen: MAPK3
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought