Order Entry
ContactUsLinkComponent
Anti-MAP2K2 Mouse Monoclonal Antibody (DL680) [Clone: 19G10.F1.E2]
Anti-MAP2K2 Mouse Monoclonal Antibody (DL680) [Clone: 19G10.F1.E2]
Catalog #: 76235-040
MEK2 N-Term MAB 100UG
Anti-MAP2K2 Mouse Monoclonal Antibody (DL680) [Clone: 19G10.F1.E2]
Catalog #: 76235-040
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Monoclonal
  • Clone:
    19G10.F1.E2
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Antigen symbol:
    MAP2K2
  • Conjugation:
    DyLight® 680
  • ELISA:
    Yes
  • Size:
    100 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Ambient
  • Purification:
    Protein A purified
  • Cat. no.:
    76235-040

Specifications

About this item

MEK2 antibodies detect the MEK2 isoform.  Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation.   This pathway also plays a key role in synaptic plasticity in the brain.  Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase.   MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain.   The MEK2 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.

Anti-MEK 2 DyLight™680 Conjugated (MOUSE) Antibody is suitable for use in Western Blotting and ELISA.  Specific conditions of reactivity should be optimized by the end user.  Expect a band of approximately 44 kDa.

Type: Primary
Antigen: MAP2K2
Clonality: Monoclonal
Clone: 19G10.F1.E2
Conjugation: DL680
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:

Customers Who Bought This Also Bought