Order Entry
ContactUsLinkComponent
Anti-KLF4 Rabbit Polyclonal Antibody
Anti-KLF4 Rabbit Polyclonal Antibody
Catalog #: 76262-956
KLF4 Polyclonal Antibody 25UL
Anti-KLF4 Rabbit Polyclonal Antibody
Catalog #: 76262-956
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    KLF4
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-956

Specifications

About this item

KLF4 belong to the Kruppel like factors/Zinc fingers C2H2-type family. This gene encodes a protein that belongs to the Kruppel family of transcription factors. KLF4 can act both as activator and as repressor. It can bind the 5-CACCC-3 core sequence, the promoter region of its own gene, and can activate its own transcription. KLF4 regulates the expression of key transcription factors during embryonic development, playing an important role in maintaining embryonic stem cells, and in preventing their differentiation. This protein is required for normal development of the barrier function of skin postnatal, maintenance of the ocular surface, function in skeletal and kidney development, and thought to control the G1-to-S transition of the cell cycle following DNA damage by mediating the tumor suppressor gene p53. KLF4 may be associated with diseases such as Venous Hemangioma, Skin Squamous Cell Carcinoma, and pediatric lymphoma. Anti-KLF4 Antibody is useful for researchers interested in Peptide hormone metabolism, Stem Cell Differentiation Pathway Research, Wnt/Notch Pathway Research, and Cancer Research.

Anti-KLF4 Antibody has been tested in Western Blot, ELISA, and Immunofluorescence. Expect a band at ~54.7 kDa in western blot using appropriate lysates. Positive control whole cell lysates used K562 and PC3 @ 1µg/mL for WB; and PC3 with PFA at 10µg/mL in IF. Although not tested, this antibody is likely functional in immunohistochemistry, flow cytometry, and immunoprecipitation.

Type: Primary
Antigen: KLF4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought