Order Entry
ContactUsLinkComponent
Anti-IL1R2 Rabbit Polyclonal Antibody
Anti-IL1R2 Rabbit Polyclonal Antibody
Catalog #: 76234-800
IL-1 Receptor Type IL Polyclonal Antibody 100UL
Anti-IL1R2 Rabbit Polyclonal Antibody
Catalog #: 76234-800
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IL1R2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    IL1R2
  • ELISA:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    100 µL
  • Western blot:
    Yes
  • Storage temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping temperature:
    Dry Ice
  • Cat. no.:
    76234-800

Specifications

About this item

Interleukin-1 Receptor Type 2 (IL-1RII) ins a non-signaling receptor for IL1A, IL1B and IL1RN and reduces IL1B activities. It serves as a decoy receptor by competitive binding to IL1B and preventing its binding to IL1R1. It also modulates cellular response through non-signaling association with IL1RAP after binding to IL1B. IL1R2 (membrane and secreted forms) preferentially binds IL1B and poorly IL1A and IL1RN. The secreted IL1R2 recruits secreted IL1RAP with high affinity; this complex formation may be the dominant mechanism for neutralization of IL1B by secreted/soluble receptors.

This antiserum against Human IL-1 Receptor Type II (IL-1RII) has been tested for use in ELISA, radioimmunoassays and immunoprecipitation.  Reactivity in other immunoassays is unknown.  IL-1RII has been termed p68 or CD121 in some laboratories.  IL-1RII has been shown to bind IL-1, IL-1a and IL-1b.

Type: Primary
Antigen: IL1R2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought