To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Rabbit
- Antigen symbol:IL1R2
- ELISA:Yes
- Size:100 µL
- Western blot:Yes
- Storage temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping temperature:Dry Ice
- Purification:Protein A purified
- Cat. no.:76234-948
Specifications
About this item
Interleukin 1 receptor type II binds to IL-1 alpha, IL-1 beta and IL1R1/IL1RA. Anti-IL-1 Receptor Type II antibody is ideal for investigators involved in Immunology and Stem Cell research.
This IgG fraction antibody of anti-Human IL-1 Receptor Type II (IL-1RII) has been tested for use in ELISA, radioimmunoassays, flow cytometry and immunoprecipitation. Reactivity in other immunoassays is unknown. IL-1RII has been termed p68 or CD121 in some laboratories. IL-1RII has been shown to bind IL-1, IL-1a and IL-1b.This product has been assayed for the ability to immunoprecipitate antigen. A dilution range of 1:400 to 1:800 is suggested for this immunoassay. For immunoprecipitation, pre-clearing with a non-specific rabbit IgG is helpful to reduce background. This product has been assayed by immunoblot using HRP Goat-anti-Rabbit IgG [H&L] (code # 611-1302) and TMB as a substrate. A working dilution range of 1:200 to 1:400 is suggested for this application. This product has been assayed by ELISA against soluble (extracellular) IL-1RII using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature. A working dilution range of 1:1,000 to 1:2000 is suggested for this product. For use in ELISA formats, this antibody is best used as the second antibody in combination with a monoclonal antibody as a capture antibody. This product has been assayed by radioimmunoassay against antigen. A dilution of 1:8,000 is suggested for this immunoassay. This product has been assayed for the ability to immunoprecipitate antigen. A dilution range of 1:400 to 1:800 is suggested for this immunoassay. For immunoprecipitation, pre-clearing with a non-specific rabbit IgG is helpful to reduce background. This product has been assayed by immunoblot using HRP Goat-anti-Rabbit IgG [H&L] (code # 611-1302) and TMB as a substrate. A working dilution range of 1:200 to 1:400 is suggested for this application. This product has been assayed by ELISA against soluble (extracellular) IL-1RII using HRP Conjugated Anti-Rabbit IgG [H&L] (Goat) (code # 611-1302) and ABTS as a substrate for 30 minutes at room temperature. A working dilution range of 1:1,000 to 1:2000 is suggested for this product. For use in ELISA formats, this antibody is best used as the second antibody in combination with a monoclonal antibody as a capture antibody. This product has been assayed by radioimmunoassay against antigen. A dilution of 1:8,000 is suggested for this immunoassay.
Type: Primary
Antigen: IL1R2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: