Order Entry
ContactUsLinkComponent
Anti-HSF1 Rabbit Polyclonal Antibody
Anti-HSF1 Rabbit Polyclonal Antibody
Catalog #: 76262-728
HSF1 Polyclonal Antibody 25UG
Anti-HSF1 Rabbit Polyclonal Antibody
Catalog #: 76262-728
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    HSF1
  • ELISA:
    Yes
  • Size:
    25 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-728

Specifications

About this item

Anti-HSF1 Antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Heat shock factor 1 (HSF1) is a highly conserved transcription factor that coordinates stress-induced transcription and directs versatile physiological processes in eukaryotes. Upon stress, HSF1 is the key transcriptional activator of chaperones, co-chaperones and ubiquitin, and also coordinates the expression of many transcriptional and translational regulators, signaling molecules and mitotic determinants. HSF1 is involved in balancing core cellular processes during stress and enables their rapid re-establishment once conditions suitable for proliferation have been restored. Importantly, HSF1 controls a distinct set of target genes in cell stress, development and cancer progression. The central role of HSF1 in diverse cellular functions is reflected in pathologies, such as neurodegenerative diseases and cancer, where an imbalance in HSF1 activity facilitates disease onset. The ability of cancer cells to harness HSF1 for metastatic progression highlights the plasticity of HSF1 in rewiring transcription and coordinating cellular processes. Anti-HSF1 antibody is thus ideal for investigators involved in cancer research and neurodegenerative diseases.

Anti-Heat Shock Factor 1 Antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band of 57 kDa in size corresponding to HSF1 by western blotting in the appropriate cell lysate or extract. Tested using positive control A431 Whole Cell Lysate p/n W09-000-361.

Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought