To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:4B3.B5.F3.B7
- Host:Mouse
- Isotype:IgG1 kappa
- Antigen symbol:HbG
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Storage temperature:Store vial at –20 °C. This product is stable at 4 °C as an undiluted liquid. For extended storage, aliquot contents and freeze at –20 °C or below Avoid cycles of freezing and thawing. Dilute only prior to immediate use.
- Shipping temperature:Dry Ice
- Purification:Protein A purified
- Cat. no.:76234-986
Specifications
About this item
HbF antibodies detect the hemoglobin gamma isoform subunit. Functional alternate hemoglobin (Hb) is a hetero tetramer composed of 2 alpha and 2 gamma subunits (alpha-2 gamma-2). Hemoglobin F is elevated in newborns, reaching adult levels by 12 months. HbF levels are increased to as much as 5% to 10% in normal pregnancy. Sickle cell disease (SCD), thalassemias and hemoglobinopathies occur when aberrant forms of hemoglobin are expressed in children and adults. Hemoglobin variants arise from mutations in the globin genes and sickle cell disease and the more benign sickle cell trait are observed in more than 100 million people globally. HbF antibody does not react other forms of Hb. This antibody is ideal for investigators involved in Cardiovascular and developmental biology research.
Anti-Hemoglobin beta F (MOUSE) antibody is suitable for use in Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 16 kDa.
Type: Primary
Antigen: HbG
Clonality: Monoclonal
Clone: 4B3.B5.F3.B7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: