Order Entry
ContactUsLinkComponent
Anti-HbB Mouse Monoclonal Antibody [Clone: 14G2.G11.F11]
Anti-HbB Mouse Monoclonal Antibody [Clone: 14G2.G11.F11]
Catalog #: 76234-980
Hemoglobin A (beta chain) MAB 25UL
Anti-HbB Mouse Monoclonal Antibody [Clone: 14G2.G11.F11]
Catalog #: 76234-980
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Monoclonal
  • Clone:
    14G2.G11.F11
  • Host:
    Mouse
  • Isotype:
    IgG2a lambda
  • Antigen symbol:
    HbB
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Protein A purified
  • Cat. no.:
    76234-980

Specifications

About this item

HbA antibodies detect the hemoglobin beta subunit wild type variant A isoform. Functional adult hemoglobin (Hb) is a hetero tetramer composed of 2 alpha and 2 beta subunits (α2β2). Common isoform variants of hemoglobin include HbA, HbS, HbC, HbF, and HbA-2. Sickle cell disease (SCD), thalassemias and hemoglobinopathies occur when aberrant forms of hemoglobin are expressed in children and adults. Globin gene mutations affect the structure and expression levels of Hb. Sickle cell disease and the more benign sickle cell trait are observed in more than 100 million people globally. Perhaps the most significant mutation is the E6V in the beta subunit and the cause of SCD, but other relevant isoforms of Hb are observed. HbA antibody cross reacts with HbA-2 but does not react other forms Hb. This antibody is ideal for investigators involved in Cardiovascular and developmental biology research.

Anti-Hemoglobin A (beta chain) (MOUSE) antibody is suitable for use in Western Blotting. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 16 kDa.

Type: Primary
Antigen: HbB
Clonality: Monoclonal
Clone: 14G2.G11.F11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a lambda
Reactivity:

Customers Who Bought This Also Bought