Order Entry
ContactUsLinkComponent
Anti-Ffar4 Rabbit Polyclonal Antibody
Anti-Ffar4 Rabbit Polyclonal Antibody
Catalog #: 76262-736
Ffar4 PAB 25UL
Anti-Ffar4 Rabbit Polyclonal Antibody
Catalog #: 76262-736
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    Ffar4
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-736

Specifications

About this item

Anti-Ffar4 Antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Free Fatty Acid 4 receptor (Ffa4 receptor or GPR120), a rhodopsin-like G protein coupled receptor (GPCR) subfamily member, is a receptor that senses specific fatty acids such as omega-3 fatty acid in fish oil or the endogenous signaling lipid, PHASA. Ffa4 receptor is enriched in lung, colon and adipose tissue but is also detected in many other tissues and cells. The activation of Ffar4 has multiple effects, including but not limited to inhibition of inflammation, improving insulin sensitivity and adipogenesis, and regulating hormone secretion from the gastro-intestinal system and pancreatic islets. Therefore, approaches that regulate FFA4 receptor activity could be developed as promising anti-diabetic and anti-inflammation drugs. GPR120 is the only fatty acid receptor that can sense lipids in adipose tissue, mature adipocytes, CD11c+ macrophages, and RAW264.7 cells making this receptor of potential importance in the prevention and treatment of metabolic and inflammatory diseases.

Anti-Ffar4 Antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band ~40-48 kDa in size corresponding to Ffar4 by western blotting in the appropriate cell lysate or extract. Tested using positive controls Raw 264.7 WCL p/n W10-001-369 and NIH/3T3 WCL p/n W10-000-358.

Type: Primary
Antigen: Ffar4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought