Order Entry
ContactUsLinkComponent
Anti-EZH1 Rabbit Polyclonal Antibody
Anti-EZH1 Rabbit Polyclonal Antibody
Catalog #: 76262-952
Ezh1 Polyclonal Antibody 25UL
Anti-EZH1 Rabbit Polyclonal Antibody
Catalog #: 76262-952
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    EZH1
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-952

Specifications

About this item

Anti-Ezh1 Antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Anti-EZH1 Antibody detects human EZH1. Polycomb group (PcG) proteins are essential epigenetic regulators in normal tissue homeostasis and are involved in transcriptional repression. The PcG members Ezh2 and Ezh1 are important determinants of embryonic stem cell identity, and the transcript levels of these histone methyltransferases are inversely correlated during development. Recent studies have shown that EZH1 also has histone H3K27 methyltransferase activity and binds to an overlapping subset of genes. EZH1 and EZH2 have different expression patterns. EZH2 is found in actively proliferating cells, whereas EZH1 expression is higher in nonproliferative adult tissues. EZH1 partially compensates for the loss of EZH2, as shown in cells lacking only Ezh2. In mice, EZH1 is a regulator of homeotic gene expression implicated in the assembly of repressive protein complexes in chromatin. Anti-EZH1 Antibody is ideal for investigators studying pathogenesis of hematological malignancies.

This affinity purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band ~86 kDa in size corresponding to Ezh1 by western blotting in the appropriate cell lysate or extract. Tested using positive control NIH/3T3 nuclear extract p/n W10-001-A74.

Type: Primary
Antigen: EZH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought