Order Entry
ContactUsLinkComponent
Anti-COL5A1 Rabbit Polyclonal Antibody (FITC)
Anti-COL5A1 Rabbit Polyclonal Antibody (FITC)
Catalog #: 76262-774
Collagen Type V PAB 50UG
Anti-COL5A1 Rabbit Polyclonal Antibody (FITC)
Catalog #: 76262-774
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    COL5A1
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    50 µg
  • Western blot:
    Yes
  • Shipping temperature:
    Ambient
  • Purification:
    immunoaffinity chromatography
  • Cat. no.:
    76262-774

Specifications

About this item

In muscle tissue, collagen serves as a major component of the endomysium. Collagen constitutes one to two percent of muscle tissue, and accounts for 6% of the weight of strong, tendinous muscles. A collagen may be defined as a protein containing sizable domain(s) of triple-helical conformation. Type IV collagen is a major macromolecular constituent of basement membranes and can be readily isolated from basement-membrane-rich tissues or highly vascularized tissues such as the placental villi. This collagen appears to be largely restricted to structures identifiable as basement membranes. In contrast, type VI collagen appears to be prevalent in several tissues even though it has been isolated largely from placental villi preparations. The extent to which type VII and VIII collagens are distributed is not known.

Anti-Collagen V Fluorescein Conjugated Antibody was assayed by immunoblot and found to be reactive against Collagen V at a dilution of 1:5,000 to 1:10,000.  This product was also assayed against 1.0 ug of Collagen V in a standard sandwich ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:4,000 to 1:8,000 of the stock concentration is suggested for this product.  For immunohistochemistry on frozen tissue sections dilute the product 1:50 to 1:200.

Type: Primary
Antigen: COL5A1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought