Order Entry
ContactUsLinkComponent
Anti-Ccnd3 Rabbit Polyclonal Antibody
Anti-Ccnd3 Rabbit Polyclonal Antibody
Catalog #: 76262-662
Cyclin D3 Polyclonal Antibody 25UL
Anti-Ccnd3 Rabbit Polyclonal Antibody
Catalog #: 76262-662
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    Ccnd3
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-662

Specifications

About this item

Anti-Cyclin D3 antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Cyclin D3 belongs to a highly conserved cyclin family, whose members are the ultimate recipients of oncogenic signals. Cyclin D3 is a key component of the cell cycle progression machinery and induces progression through the G1 phase of the cell cycle. Cyclin D3 is expressed in nearly all proliferating cells, and shows the most broad expression pattern of all three D-type (D1-D3) cyclins. Cyclin D3 is encoded from the 6p21 chromosome region and the protein is predominantly localized in the nucleus. Once induced, cyclin D3 binds and activates its associated cyclin-dependent kinases CDK4 and CDK6. Amplification of the cyclin D3 gene and overexpression of cyclin D3 protein is seen in several human cancers. A large number of human malignancies contain lesions in pathways impacting on cyclin D3. Abnormal expression of Cyclin D3 is believed to be a driving force in several human cancers. A possible role for cyclin D3 in the malignancies of the lymphoid system is suggested by the observations that cyclin D3 gene is rearranged in several neoplastic diseases such as diffuse large B cell lymphomas or multiple myelomas. Anti-Cyclin D3 is ideal for researchers interested in Cancer Research and Immunology research.

This affinity purified antibody has been tested for use in ELISA, immunohistochemistry, flow cytometry, and by immunoprecipitation. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 32.4 kDa in size corresponding to Cyclin D3 protein by western blotting in the appropriate stimulated tissue or cell lysate or extract. This reagent reacts equally to phosphorylated and non-phosphorylated Cyclin D3 by ELISA and western blotting.

Type: Primary
Antigen: Ccnd3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought