Order Entry
ContactUsLinkComponent
Anti-ASAP1 Rabbit Polyclonal Antibody
Anti-ASAP1 Rabbit Polyclonal Antibody
Catalog #: 76262-752
ASAP1 Polyclonal Antibody 25UL
Anti-ASAP1 Rabbit Polyclonal Antibody
Catalog #: 76262-752
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    ASAP1
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-752

Specifications

About this item

This antibody is designed, produced, and validated as part of a collaboration between Rockland and the National Cancer Institute (NCI). ASAP1 (ArfGAP with SH3 Domain, Ankyrin Repeat and PH Domain 1) is a Protein Coding gene. This gene encodes an ADP-ribosylation factor (ARF) GTPase-activating protein. The GTPase-activating activity is stimulated by phosphatidylinositol 4,5-biphosphate (PIP2), and is greater towards ARF1 and ARF5, and lesser for ARF6. This gene maybe involved in regulation of membrane trafficking with cell growth or actin cytoskeleton remodeling by binding to both SRC and PIP2. ASAP1 may function as a signal transduction protein involved in the differentiation of fibroblasts into adipocytes and possibly other cell types (By similarity) and plays a role in ciliogenesis. Anti-ASAP1 is useful for researchers interested in GTPase activities and EGFR1 Signaling Pathway.

Anti-ASAP1 Antibody has been tested for use in ELISA and Western Blot. Expect a band approximately ~125kDa using positive control lysates over-expressing ASAP1, or other specific lysates or tissues. Although not tested, this antibody could be useful in Immunofluorescence or Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: ASAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought