Order Entry
ContactUsLinkComponent
Anti-ALG6 Rabbit Polyclonal Antibody
Anti-ALG6 Rabbit Polyclonal Antibody
Catalog #: 76234-750
ALG6 Polyclonal Antibody 100UL
Anti-ALG6 Rabbit Polyclonal Antibody
Catalog #: 76234-750
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    ALG6
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    100 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    multi-step process which includes delipidation and salt fractionation
  • Cat. no.:
    76234-750

Specifications

About this item

Anti-ALG6 antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase is an enzyme that is encoded by the ALG6 gene in humans. This gene encodes ALG6/ALG8 member of glucosyltransferase family. The encoded protein catalyzes the addition of the first glucose residue to the growing lipid-linked oligosaccharide precursor of N-linked glycosylation. Mutations in this gene are associated with congenital disorders of glycosylation type. Anti-ALG6 is ideal for researchers interested in Infectious disease, especially HIV and hepatitis virus infections. Anti-ALG6 antibody is applicable to methods for diagnosis and, in particular, to methods for diagnosing infections using biomarkers targeting exosomes secreted in bodily fluids.

Anti-ALG6 antibody has been tested in ELISA, western blot, and immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 58kDa in size corresponding to ALG6 protein by Western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: ALG6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought