Order Entry
ContactUsLinkComponent
Anti-Abi1 Rabbit Polyclonal Antibody
Anti-Abi1 Rabbit Polyclonal Antibody
Catalog #: 76262-940
Abi1 Polyclonal Antibody 25UL
Anti-Abi1 Rabbit Polyclonal Antibody
Catalog #: 76262-940
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Antigen symbol:
    Abi1
  • ELISA:
    Yes
  • Size:
    25 µL
  • Western blot:
    Yes
  • Shipping temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. no.:
    76262-940

Specifications

About this item

Anti-Abi1 Antibody was designed, produced, and validated as part of the Joy Cappel Young Investigator Award (JCYIA). Abi1 gene has been suggested to have a role in inhibiting cell growth and encodes an adaptor protein in the Abelson-interactor family. The protein is involved in signal transduction, actin regulation and cytoskeletal remodeling. Studies suggest that Abi1 is a member of the WAVE complex and a dysfunctional complex may allow for the progression of prostate cancer. Anti-Abi1 is ideal for researchers interested in melanoma, colon and breast cancer and other cancer antibody research.

This affinity purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band ~ 65 kDa in size corresponding to Abi1 by western blotting in the appropriate cell lysate or extract.

Type: Primary
Antigen: Abi1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Customers Who Bought This Also Bought