N-Methyl-2-pyrrolidone (NMP) ≥99.8%, Finyte® for microelectronic, J.T.Baker®
Catalog #: CAJT6337-5
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 872-50-4
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, pressure bottom
- Size:4 L
- Cat. no.:CAJT6337-5
- Supplier No.:6337-05
Specifications
Specification Test Results
- For Microelectronic Use:
- Assay (C₅H₉NO):≥99.8 %
- Color (APHA):≤30
- Water (H₂O)(by Karl Fischer titrn):≤300 ppm
- Free Amines (as CH₃NH₂):≤50 ppm
- Chloride (Cl):≤0.5 ppm
- Phosphate (PO₄):≤1 ppm
- Trace Impurities - Aluminum (Al):≤10.0 ppb
- Arsenic and Antimony (as As):≤10.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Beryllium (Be):≤10.0 ppb
- Trace Impurities - Bismuth (Bi):≤10.0 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤10.0 ppb
- Trace Impurities - Chromium (Cr):≤10.0 ppb
- Trace Impurities - Cobalt (Co):≤10.0 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤10.0 ppb
- Trace Impurities - Germanium (Ge):≤10.0 ppb
- Trace Impurities - Gold (Au):≤10.0 ppb
- Trace Impurities - Iron (Fe):≤10.0 ppb
- Trace Impurities - Lead (Pb):≤10.0 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤10.0 ppb
- Trace Impurities - Manganese (Mn):≤10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤10.0 ppb
- Trace Impurities - Nickel (Ni):≤10.0 ppb
- Trace Impurities - Niobium (Nb):≤10.0 ppb
- Trace Impurities - Potassium (K):≤10.0 ppb
- Trace Impurities - Silver (Ag):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤10.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤10.0 ppb
- Trace Impurities - Thallium (Tl):≤10.0 ppb
- Trace Impurities - Tin (Sn):≤10.0 ppb
- Trace Impurities - Titanium (Ti):≤10.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤10.0 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
- Particle Count - 0.3 µm and greater (Rion KS42AF):
- Particle Count - 0.5 µm and greater (Rion KS42AF):≤100 par/ml
- Particle Count - 1.0 µm and greater (Rion KS42AF):≤10 par/ml
Specification Test Results
About this item
Synonyms: 1-Methyl-2-pyrrolidinone,1-Methyl-2-pyrrolidone
:Storage Temperature: Ambient
Density:1.03 g/cm³
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Management of Change (MOC) category = R
Flash Pt:96.00 °C
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: -24 °C
:Flash Pt:
:Flash Pt: 96.00 °C
:M.W. 99.13 g/mol
:-24
:202
:96.00 °C
:GHS07
:GHS08
Storage Temperature:Ambient
:M.O.C. Category R
:12,06197
Formula:C₅H₉NO
MW:99.13 g/mol
MDL Number:MFCD00003193
CAS Number:872-50-4
Customers Who Bought This Also Bought