ContactUsLinkComponent
Rubber Stopper for Serum Bottles and Vials, Ace Glass Incorporated
Rubber Stopper for Serum Bottles and Vials, Ace Glass Incorporated
Catalog #: 76291-836
Supplier:  Ace Glass
Stopper Snap-On Thin Flange 13X20Mm CS1
Rubber Stopper for Serum Bottles and Vials, Ace Glass Incorporated
Catalog #: 76291-836
Supplier:  Ace Glass
Specifications
  • Description:
    Snap-On Rubber Stopper
  • Cat. no.:
    76291-836
  • I.D.×O.D.:
    13×20 mm
Specifications
About this item
Gray, thin, flange-style chlorobutyl rubber stopper fits all 20 mm serum bottles and serum vials.
Customers Who Bought This Also Bought