(±)-1,2-Propanediol ≥99.5% (by GC) USP, FCC, Macron Fine Chemicals™
Catalog #: CAMK626319
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 57-55-6
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Steel drum, phenolic lined
- Size:20 L
- Cat. no.:CAMK626319
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets F.C.C. Requirements:
- Meets U.S.P Requirements:
- Food GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Bulk Pharmaceutical Chemical:
- USP - Assay (CH₃CHOHCH₂OH) (by GC):≥ 99.5 %
- USP - Identificaton A (IR):Passes Test
- USP - Identification B - Diethylene Glycol:≤ 0.10 %
- USP - Identification B - Ethylene Glycol:≤ 0.10 %
- USP - Identification C:Passes Test
- USP - Acidity:Passes Test
- USP - Water (H₂O)(by Karl Fischer titrn):≤ 0.2 %
- USP - Residue on Ignition:≤ 0.007 %
- USP - Chloride (Cl):≤ 0.007 %
- USP - Sulfate (SO₄):≤ 0.006 %
- FCC - Assay (CH₃CHOHCH₂OH) (by GC):≥ 99.5 %
- FCC - Acidity:Passes Test
- FCC - Identificaton A (IR):Passes Test
- FCC - Lead (Pb):≤ 1 mg/kg
- FCC - Residue on Ignition:≤ 0.007 %
- FCC - Specific Gravity at 25°/25°C:1.035 - 1.037
- FCC - Water (H₂O)(by Karl Fischer titrn):≤ 0.20 %
- FCC - Distilling Range(Lower):185 - 189 °C
- FCC - Distilling Range(Upper):185 - 189 °C
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Metallic Residues: No metal catalysts or metal reagents, as defined by:
- EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of:
- this material.:
Specification Test Results
About this item
Synonyms: Propylene glycol,1,2-Propanediol,(±)-1,2-Propylene glycol,1,2-Dihydroxypropane,(±)-1
:Storage Temperature: Ambient
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = R
Flash Pt:99 °C
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: -59 °C
:Flash Pt:
:Flash Pt: 99 °C
:M.W. 76.1 g/mol
:-59
:187.6
:99 °C
MW:76.1 g/mol
Storage Temperature:Ambient
:M.O.C. Category R
:12,08040
Formula:CH₃CH(OH)CH₂OH
MDL Number:MFCD00064272
CAS Number:57-55-6
Customers Who Bought This Also Bought