Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular USP, ACS, J.T.Baker®
Catalog #: CAJT2262-20
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 50-99-7
MOC: Regulated
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PE drum, open head
- Size:100 kg
- Cat. no.:CAJT2262-20
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets ACS Reagent Chemical Requirements:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C: Water (H₂O):≤1.0 %
- USP - Assay (Anhydrous basis):97.5 - 102.0 %
- USP - Related substances: Maltose and isomaltose:≤0.4 %
- USP - Dextrin:Passes Test
- USP - Related substances: Maltotriose:≤0.2 %
- USP - Related substances: Fructose:≤0.15 %
- USP - Related substances: Unspecified impurities, each:≤0.10 %
- USP - Related substances: Total Impurities:≤0.5 %
- USP - Color and Clarity of Solution:Passes Test
- USP - Soluble Starch, Sulfites:≤15 ppm
- USP - Conductivity (uS cm-1):≤20 µS/cm
- Chloride (Cl):≤0.01 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Insoluble Matter:≤0.005 %
- Loss on Drying at 105°C:≤0.2 %
- Residue after Ignition:≤0.015 %
- Solution in Water (50%)(APHA):≤20
- Specific Rotation [Å]²⁵ ^D (+):52.5 - 53.0 Degree
- Starch:Passes Test
- Sulfate and Sulfite (as SO₄):≤0.005 %
- Titrable Acid (meq/g):≤0.002
- Trace Impurities - Iron (Fe):≤5 ppm
- Appearance (white, granular powder):
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular USP, ACS:
Specification Test Results
About this item
Synonyms: Corn sugar,Glucose,D-(+)-Dextrose,(2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal,Glucolin,Dextrose
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97,5-102,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 146 °C
:Flash Pt:
:Flash Pt:
:M.W. 180.16 g/mol
:Dextrose
:146
:12,04467
:14,04459
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MW:180.16 g/mol
MDL Number:MFCD00148912
Customers Who Bought This Also Bought