Borate Buffered Saline (BBS), Rockland Immunochemicals
Catalog #: 76237-550
Supplier: Rockland Immunochemical
CAS Number:
Specifications
- Description:Borate Buffered Saline (BBS)
- Size:1 L
- Cat. no.:76237-550
Specifications
About this item
Borate buffered saline (BBS) is commonly used in coating procedures for ELISA.
- Borate buffered saline - BBS
- Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
- Tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria
BBS can also be used as a component in blocking solutions for Western blotting. The buffer can be used as a diluent in biological samples and for various applications within biology and biochemistry to maintain the pH.
Application Note: BBS is isotonic and has a strong bactericidal effect. It can be used to dilute solutions for coating procedures. Additives such as Tween 20 and Blotto can be used to add functionality as a washing buffer or blocking buffer.
Customers Who Bought This Also Bought