L(+)-Valine 98.5-101.5% (dried basis) USP, FCC, Multi-Compendial, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: H-Val-OH,Valine
Formula:C₅H₁₁NO₂
MW:117.15 g/mol
Density:1.23 g/cm³
MDL Number:MFCD00064220
CAS Number:72-18-4
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5-101,5
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 86 °C
:Flash Pt:
:Flash Pt:
:M.W. 117.15 g/mol
:86
:217
:12,10046
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets B.P. Chemical Specifications:
- GMP Manufactured Product:
- Meets E.P. Chemical Specifications:
- Meets J.P. Chemical Specifications:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (C₅H₁₁NO₂) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²⁵ ^D (+):26.6 - 28.8 Degree
- USP - pH:5.5 - 7.0
- USP - Loss on Drying at 105°C:≤0.3 %
- USP - Residue on Ignition:≤0.1 %
- USP - Chloride (Cl):≤0.05 %
- USP - Sulfate (SO₄):≤0.03 %
- USP - Iron (Fe):≤0.003 %
- USP - Related Compounds - Individual Impurities:≤0.5 %
- USP - Related Compounds - Total Impurities:≤2.0 %
- EP/BP - Assay (C₅H₁₁NO₂) (dried basis):98.5 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Specific Rotation [Å]²º^D (+):26.5 - 29.0 Degree
- EP/BP-Ninhydrin-Positive Substances-Impurity B:≤0.4 %
- EP/BP-Ninhydrin-Positive Substances-Each:≤0.2 %
- EP/BP-Ninhydrin-Positive Substances-Total Impurities:≤1.0 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Sulfate (SO₄):≤300 ppm
- EP/BP - Ammonium (NH₄):≤0.02 %
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Loss on Drying:≤0.5 %
- EP/BP - Ash (sulfated):≤0.1 %
- JP - Assay (C₅H₁₁NO₂) (dried basis):98.5 - 101.0 %
- JP - Identification:Passes Test
- JP - Optical Rotation (+):26.5 - 29.0 Degree
- JP - pH:5.5 - 6.5
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.021 %
- JP - Sulfate (SO₄):≤0.028 %
- JP - Ammonium (NH₄):≤0.02 %
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Related Substances:Passes Test
- JP - Loss on Drying at 105°C:≤0.30 %
- JP - Residue on Ignition:≤0.10 %
- Endotoxin Concentration, IU/mg, For Information Only:
- Only Class 2 Solvents (Methanol) are likely to be present. All are:
- below Option 1 limits.:
- Metallic Residues: No metal catalysts or metal reagents, as defined by:
- EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of:
- this material.:
Specification Test Results
Customers Who Bought This Also Bought