Kimble® Disposable NMR Tubes, 10 mm, Flat Bottom, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
All tubes feature a 10 mm Ext.Ø, and have a wall thickness of 0.381 mm.
- Flat bottom design
- Caps not included
Certifications: Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
Customers Who Bought This Also Bought