Ponceau S solution
Catalog #: 76236-520
Supplier: Rockland Immunochemical
CAS Number: 6226-79-5
Specifications
- Size:500 ml
- Cat. no.:76236-520
Specifications
About this item
Synonyms: 3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt,3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt,CI 27195,3-Hydroxy-4-((2-sulfo-4-((4-sulfophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulfonic acid, tetrasodium salt,Ponceau,3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 760.56 g/mol
Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
MW:760.56 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00003892
CAS Number:6226-79-5
Customers Who Bought This Also Bought