Order Entry
ContactUsLinkComponent
Rat DKK1 ELISA Kit, Rockland Immunochemicals
Rat DKK1 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-526
CAS Number:  
Rat DKK1 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-526
CAS Number:  

Specifications

  • Description:
    Rat DKK1 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-526

Specifications

About this item

Rat DKK1 AccuSignal ELISA Kit

  • Natural and recombinant rat DKK1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S23-H270

Dickkopf-related protein 1 is a protein that in humans is encoded by the DKK1 gene, which was mapped to chromosome 10q11.2. This gene encodes a protein that is a member of the dickkopf family. It is a secreted protein with two cysteine rich regions and is involved in embryonic development through its inhibition of the WNT signaling pathway. Elevated levels of DKK1 in bone marrow, plasma and peripheral blood is associated with the presence of osteolytic bone lesions in patients with multiple myeloma.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 3000pg/ml, 1500pg/ml, 750pg/ml, 419.pg/ml, 187.5pg/ml, 93.8pg/ml, 46.9pg/ml rat DKK1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernatants, serum or plasma (heparin, EDTA) to each empty well. We recommend that each rat DKK1 standard solution and each sample is measured in duplicate.

Customers Who Bought This Also Bought