Order Entry
ContactUsLinkComponent
Human ERBB3 - Her3 ELISA Kit, Rockland Immunochemicals
Human ERBB3 - Her3 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-632
CAS Number:  
Human ERBB3 - Her3 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-632
CAS Number:  

Specifications

  • Reactivity:
    Human
  • Description:
    Human ERBB3 - Her3 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-632

Specifications

About this item

Human ERBB3 - Her3 AccuSignal ELISA Kit

  • Natural and recombinant human ERBB3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S20-T643

Receptor tyrosine-protein kinase erbB-3, also known as HER3 (human epidermal growth factor receptor 3), is a membrane bound protein that in humans is encoded by the ERBB3 gene. ErbB3 has been shown to bind the ligands heregulin and NRG-2. Ligand binding causes a change in conformation that allows for dimerization, phosphorylation, and activation of signal transduction. ErbB3 can heterodimerize with any of the other three ErbB family members. The theoretical ErbB3 homodimer would be non-functional because the kinase-impaired protein requires transphosporylation by its binding partner to be active. Unlike the other ErbB receptor tyrosine kinase family members which are activated through autophosphorylation upon ligand binding, ErbB3 is found to be kinase impaired, having only 1/1000th the autophosphorylation activity of EGFR and no ability to phosphorylate other proteins. Therefore, ErbB3 must act as anallosteric activator.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human ERBB3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human ERBB3 standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought