Order Entry
ContactUsLinkComponent
Mouse Epiregulin ELISA Kit, Rockland Immunochemicals
Mouse Epiregulin ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-630
CAS Number:  
Mouse Epiregulin ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-630
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Reactivity:
    Mouse
  • Description:
    Mouse Epiregulin ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-630

Specifications

About this item

Mouse Epiregulin AccuSignal ELISA Kit

  • Natural and recombinant mouse Epiregulin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: V53-L101

Epiregulin (EPR) is a protein that in humans is encoded by the EREG gene. It is a member of the epidermal growth factor family. Epiregulin can function as a ligand of epidermal growth factor receptor (EGFR), as well as a ligand of most members of the ERBB (v-erb-b2 oncogene homolog) family of tyrosine-kinase receptors. The secondary structure at the C-terminus epiregulin is different from other epidermal growth factor family ligands because of the lack of hydrogen bonds. The structural difference at the C-terminus may provide an explanation for the reduced binding affinity of epiregulin to the ERBB receptors.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 8000pg/ml, 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml mouse Epiregulin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse Epiregulin standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought