Specifications
- Reactivity:Human
- Description:Human LOXL2 ELISA Kit
- Size:1 Kit
- Shipping temperature:Wet Ice
- Cat. no.:76236-232
Specifications
About this item
Human LOXL2 AccuSignal ELISA Kit
- Natural and recombinant human LOXL2.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Q26-Q774
Lysyl oxidase homolog 2 is an enzyme that in humans is encoded by the LOXL2 gene. This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family. LOXL2 can also crosslink collagen type IV and hence influence the sprouting of new blood vessels.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human LOXL2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, cell lysates, tissue homogenates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human LOXL2 standard solution and each sample be measured in duplicate.