Order Entry
ContactUsLinkComponent
Rat Neuropilin-1 ELISA Kit, Rockland Immunochemicals
Rat Neuropilin-1 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-492
CAS Number:  
Rat Neuropilin-1 ELISA Kit, Rockland Immunochemicals
Catalog #: 76237-492
CAS Number:  

Specifications

  • Description:
    Rat Neuropilin-1 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76237-492

Specifications

About this item

Rat Neuropilin-1 AccuSignal ELISA Kit

  • Natural and recombinant rat Neuropilin-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: F22-D854

NRP1(Neuropilin 1) also known as NP1, NRP, BDCA4 or VEGF165R, is a membrane-bound coreceptor to a tyrosine kinase receptor for both vascular endothelial growth factor (VEGF) and semaphorin family members. NRP1 plays versatile roles in angiogenesis, axon guidance, cell survival, migration, and invasion. By somatic cell hybrid analysis, the NRP1 gene was mapped to chromosome 10. NRP1 bounds PGF1 with lower affinity. NRP1-mediated interactions are a necessary element in the initiation of the primary immune response and offer another example, like that of agrin, of a molecule shared by neurologic and immunologic synapses. After T-cell contact with DC, T-cell NRP1 colocalized with CD3 in the immunologic synapse and, sometimes, also at the opposite pole of the T cell. Soluble NRP1 interacts in a homophilic fashion with NRP1 on both DC and T cells, and this binding can be inhibited by blocking antibodies to NRP1. Furthermore, selective NRP1 inhibition in this model suppressed neovascular formation substantially.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml rat Neuropilin-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each rat Neuropilin-1 standard solution and each sample is measured in duplicate.

Customers Who Bought This Also Bought