Order Entry
ContactUsLinkComponent
Human TGFBR2 ELISA Kit, Rockland Immunochemicals
Human TGFBR2 ELISA Kit, Rockland Immunochemicals
Catalog #: 76236-174
CAS Number:  
Human TGFBR2 ELISA Kit, Rockland Immunochemicals
Catalog #: 76236-174
CAS Number:  

Specifications

  • Reactivity:
    Human
  • Description:
    Human TGFBR2 ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76236-174

Specifications

About this item

Human TGFBR2 AccuSignal ELISA Kit

  • Natural and recombinant human TGFBR2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: I24-D159

TGFBR2(transforming growth factor, beta receptor II (70/80kDa)), also known as TGF-beta receptor type-2, TGFR-2, TGF-beta type II receptor, Transforming growth factor-beta receptor type II( TGF-beta receptor type II, TbetaR-II), is a member of the Ser/Thr protein kinase family and the TGFB receptor subfamily. A TGFBR2 cDNA encoding a deduced 565-amino acid protein with a calculated molecular mass of approximately 60 kD in length. The encoded protein is a transmembrane protein that has a protein kinase domain, forms a heterodimeric complex with another receptor protein, and binds TGF-beta. This receptor/ligand complex phosphorylates proteins, which then enter the nucleus and regulate the transcription of a subset of genes related to cell proliferation. Mutations in this gene have been associated with Marfan syndrome, Loeys-Deitz aortic aneurysm syndrome, Osler-Weber-Rendu syndrome, and the development of various types of tumors. Alternatively spliced transcript variants encoding different informs have been cha...

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human TGFBR2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human TGFBR2 standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought