Order Entry
ContactUsLinkComponent
Rat RANK ELISA Kit, Rockland Immunochemicals
Rat RANK ELISA Kit, Rockland Immunochemicals
Catalog #: 76235-978
CAS Number:  
Rat RANK ELISA Kit, Rockland Immunochemicals
Catalog #: 76235-978
CAS Number:  

Specifications

  • Description:
    Rat RANK ELISA Kit
  • Size:
    1 Kit
  • Shipping temperature:
    Wet Ice
  • Cat. no.:
    76235-978

Specifications

About this item

Rat RANK AccuSignal ELISA Kit

  • Natural and recombinant rat RANK.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q30-P213

Receptor Activator of Nuclear Factor kB (RANK), also known as TRANCE Receptor, is a type I membrane protein that is expressed on the surface of osteoclasts and is involved in their activation upon ligand binding. RANK is a recently described TNF receptor family member, and its ligand, RANKL, promote survival of dendritic cells and differentiation of osteoclasts. RANK contains 383 amino acids in its intracellular domain(residues 234-616), which contain three putative TRAF-binding domains(termed I, II, and III). RANK interacts with various TRAFs through distinct motifs and activates NF-kappaB via a novel TRAF6 interaction motif, which then activates NIK, thus leading to NF-kappaB activation, whereas RANK most likely activates JNK through a TRAF2-interacting region in RANK. The standard in this kit is recombinant human RANK with the sequence of Q29-G213 aa. It is a dipolymer which compose of two chains, and the molecular weight of each is 48kda.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml rat RANK standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates or serum to each empty well. It is recommended that each rat RANK standard solution and each sample be measured in duplicate.

Customers Who Bought This Also Bought